missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phloxine B, 85%, pure, high purity, Biological stain

84.62 € - 218.09 €

Chemical Identifiers

CAS 18472-87-2
Molecular Formula C20H2Br4Cl4Na2O5
MDL Number MFCD00005061
Synonym Acid Red 92, C.I. 45410
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10133730
View Documents
Thermo Scientific Chemicals
189470050
5 g Glass bottle
84.62 €
5g
In Stock
to see stock.
Add to basket
 
10501641
View Documents
Thermo Scientific Chemicals
189470250
25 g Glass bottle
218.09 €
25g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18472-87-2
C20H2Br4Cl4Na2O5
MFCD00005061
Acid Red 92, C.I. 45410

Specifications

875 to 1300 (at lambda max.)
1.38 to 2.24
85%
MFCD00005061
15, 7440
Authentic
829.64
85%
Brown to Brown/Red
Phloxine B
18472-87-2
80% min. (on anhydrous substance)
C20H2Br4Cl4Na2O5
19, IV, 2921
Acid Red 92, C.I. 45410
546 to 550nm (in 50:50 METHS:water)
10% max. (105°C)
Pure
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.