Learn More
BD Phoenix™ ID Broth
Used to make a heavy suspension of a selected colony for processing in the Phoenix™ automated identification and susceptibility testing system. BD Phoenix™ ID Broth is packaged in 100 pre-measured bottles of prepared solution.
Brand:  BD 246001
Product Code. 10686925
  • 197.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Alert:
prepared, ready to use, bottle, premeasured, automated, identification, susceptibility, testing, heavy, suspension, innoculate
Description
Description
BD Phoenix™ Automated Identification and Susceptibility Testing System
  • Rapid, accurate and reliable detection of known and emerging antimicrobial resistance.
  • Workflow efficiency utilizing automated nephelometry resulting in a standardized isolate inoculum and a reduction of technologist labor.
  • State-of-the-art data management to monitor, analyze and communicate actionable results directly to laboratorians and clinicians.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microbiology Media
Phoenix™ automated identification and susceptibility testing system
100Pack
Preparing a heavy suspension of a colony
Pre-filled bottles