missing translation for 'onlineSavingsMsg'
Learn More
Phosgene iminium chloride, 98%, Thermo Scientific Chemicals Product Code.: 10114430

Phosgene iminium chloride, 98%, Thermo Scientific Chemicals

Product Code. 10114430
100 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10114430

Brand: Thermo Scientific Acros 172211000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33842-02-3
Molecular Formula C3H6Cl3N
Molecular Weight (g/mol) 162.44
MDL Number MFCD00011870
InChI Key NRNFKRFWZQQDMD-UHFFFAOYSA-M
Synonym dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chlorideShow MoreShow Less
PubChem CID 2723945
SMILES [Cl-].CN(C)[C+](Cl)Cl

Specifications

Melting Point 181°C to 185°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula Cl2C=N(CH3)2Cl
Quantity 100 g
Fieser 04,135; 05,195; 06,170; 08,156
Merck Index 15, 10174
Solubility Information Solubility in water: hydrolysis
IUPAC Name dichloromethylidene(dimethyl)azanium;chloride
Formula Weight 162.45
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phosgene iminium chloride, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466950 Certificate TypeCertificate of Analysis Date24/10/2024 Catalog Number172210250,172211000
Lot NumberA0416557 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number172210250,172211000
Lot NumberA0392711 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog Number172210250,172211000,172210050
Lot NumberA0424735 Certificate TypeCertificate of Analysis Date27/11/2023 Catalog Number172210250,172211000,172210050
Lot NumberA0386833 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number172210250,172211000,172210050
compliance-icons compliance-icons
Product Identifier
  • Phosgene iminium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosgene iminium chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.