missing translation for 'onlineSavingsMsg'
Learn More
Phosphate buffer solution, pH 6.8 EU Pharmacopoeia, Fisher Chemical™ Product Code.: 12950704

Phosphate buffer solution, pH 6.8 EU Pharmacopoeia, Fisher Chemical™

Product Code. 12950704
1000 mL
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy

Product Code. 12950704

Brand: Fisher Chemical SOL 061 1L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 7732-18-5
pH 6.8
Quantity 1000 mL
Packaging Bottle
Grade Eu. Ph. Reference
Chemical Name or Material Phosphate buffer solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1089189 Certificate TypeCertificate of Analysis Date05/03/2025 Catalog NumberSOL 061 1L
Lot Number1033087 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberSOL 061 1L
Lot Number1020701 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberSOL 061 1L
Lot Number1034667 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberSOL 061 1L
Lot Number1048739 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberSOL 061 1L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphate buffer solution, pH 6.8 EU Pharmacopoeia, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.