missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), pH 7.4, for Western blot

For Western blot washing

59.70 € - 171.86 €

Specifications

Color Colorless
Dilution 10 X
Product Type Phosphate Buffered Saline (PBS)
For Use With (Application) For Western blot washing.
pH 7.4
View More Specs

Products 3
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
15480147
View Documents
Thermo Scientific Chemicals
J60801.K3
2 L
93.06 €
2 L
Please to purchase this item. Need a web account? Register with us today!
15490147
View Documents
Thermo Scientific Chemicals
J60801.K7
4 L
171.86 €
4 L
Please to purchase this item. Need a web account? Register with us today!
15470147
View Documents
Thermo Scientific Chemicals
J60801.K2
1 L
59.70 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. It is associated with ethylenediaminetetraacetic acid (EDTA) and used to disengage attached and clumped cells. It also helps to maintain a constant pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Colorless
Phosphate Buffered Saline (PBS)
7.4
10 X
For Western blot washing.
Liquid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.