missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL, Thermo Scientific Chemicals
139.00 € - 295.00 €
Chemical Identifiers
CAS | 7558-80-7 |
---|---|
Molecular Formula | H2NaO4P |
Molecular Weight (g/mol) | 119.98 |
MDL Number | MFCD00003527,MFCD00146206 |
InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
Synonym | Phosphate, Ion chromatography standard solution |
PubChem CID | 23672064 |
ChEBI | CHEBI:37585 |
IUPAC Name | sodium;dihydrogen phosphate |
SMILES | [Na+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363647
|
Thermo Scientific Alfa Aesar
035548.AE |
100 mL |
139.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475010
|
Thermo Scientific Alfa Aesar
035548.AP |
500 mL |
295.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
23672064 | |
sodium;dihydrogen phosphate |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
Phosphate, Ion chromatography standard solution | |
CHEBI:37585 | |
[Na+].OP(O)([O-])=O |
Specifications
PO4-3 1000μg/mL | |
H2NaO4P | |
Phosphate, Ion chromatography standard solution | |
[Na+].OP(O)([O-])=O | |
119.98 | |
CHEBI:37585 | |
Liquid |
7558-80-7 | |
MFCD00003527,MFCD00146206 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate | |
23672064 | |
Miscible with water | |
Phosphate |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only