missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Phosphate Ortho/Total Cuvette Tests

Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Officially approved for consent limits. Environmentally friendly due to less chemicals.

Brand:  Hach Lange™ LCK349


 View more versions of this product

Product Code. 10000234

  • 220.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Phosphate (Ortho/Total) cuvette test 0.05-1.5 mg/L PO4-P
  • For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
  • ISO 6878-1-1986, DIN 38405 D11-4 certifications
  • Shelf life of 23 months from production date
  • Storage conditions of 15 - 25 °C
  • Measuring ranges of 0.05 - 1.5 mg/L PO4-P and 0.15 - 4.5 mg/L PO4
  • Precise and reliable measurement values
  • Convenient and error-free dosing of the reagents
Specifications

Specifications

25
15°C to 25°C
Phosphate Ortho/Total Cuvette Test
0.05 to 1.5mg/L PO4-P
Phosphormolybdenum Blue
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phosphat/Phosphate, Sample cuvette

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 3

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid, Sodium peroxidisulfate,Sodium metaborate, Lithium sulfate monohydrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.