missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
51.20 € - 107.00 €
Chemical Identifiers
CAS | 7558-80-7 |
---|---|
Molecular Weight (g/mol) | 119.98 |
InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
SMILES | [Na+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12912185
|
Fisher Chemical
A 038 100 |
100 mL | Bottle |
51.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12922185
|
Fisher Chemical
A 038 250 |
250 mL | Bottle |
73.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12932185
|
Fisher Chemical
A 038 500 |
500 mL | Bottle |
107.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
[Na+].OP(O)([O-])=O |
Specifications
1g/L | |
Single-component Standard Solution | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium dihydrogen phosphate | |
H2O | |
Phosphate |
7558-80-7 | |
PO43- | |
[Na+].OP(O)([O-])=O | |
119.98 | |
Liquid |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)