missing translation for 'onlineSavingsMsg'
Learn More

Phospho-ATR (Thr1989) Recombinant Rabbit Monoclonal Antibody (PS01-18), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen MA545973

Product Code. 17804594

  • 447.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Predicted band size: 301 kDa Positive Control: HeLa cell lysate, HeLa treated with Hydroxyurea cell lysate, HeLa treated with 4mM Hydroxyurea for 20 hours. Subcellular Location: Chromosome, Nucleus. Sequence Similarities: No Cross with Mouse and Rat. Tissue Specificity: Ubiquitous, with highest expression in testis. Isoform 2 is found in pancreas, placenta and liver but not in heart, testis and ovary.

The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-ATR (Thr1989)
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide
Q13535
ATR
Synthetic peptide.
100 μL
Primary
Human
Liquid
Immunocytochemistry, Western Blot
PS01-18
Unconjugated
ATR
2310008J16Rik; 2810405N18Rik; ataxia telangiectasia and Rad3 related; ataxia telangiectasia and Rad3-related protein; ATR; ATR serine/threonine kinase; EC 2.7.11.1; FCTCS; FRAP-related protein 1; FRAP-related protein-1; FRP1; I79_000761; kinase ATR; MEC1; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; SCKL; SCKL1; Serine/threonine-protein kinase ATR; TEM8
Rabbit
Protein A
RUO
545
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers