missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-SF3B1 (Thr313) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5104648
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of SF3B1 only when phosphorylated at Thr313.
This gene encodes subunit 1 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex. The splicing factor 3b/3a complex binds pre-mRNA upstream of the intron's branch site in a sequence independent manner and may anchor the U2 snRNP to the pre-mRNA. Splicing factor 3b is also a component of the minor U12-type spliceosome. The carboxy-terminal two-thirds of subunit 1 have 22 non-identical, tandem HEAT repeats that form rod-like, helical structures. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
Phospho-SF3B1 (Thr313) | |
Polyclonal | |
Unconjugated | |
SF3B1 | |
155kDa; 2810001M05Rik; AA409119; Hsh155; LOW QUALITY PROTEIN: splicing factor 3B subunit 1; MDS; pre-mRNA processing 10; pre-mRNA splicing factor SF3b, 155 kDa subunit; pre-mRNA-splicing factor SF3b 155 kDa subunit; PRP10; PRPF10; SAP 155; SAP155; SF3B1; SF3b155; spliceosome-associated protein 155; Splicing factor 3B subunit 1; splicing factor 3b subunit 1, 155kDa; splicing factor 3b, subunit 1; splicing factor 3b, subunit 1, 155 kDa; splicing factor 3b, subunit 1, 155kD; splicing factor 3b, subunit 1, 155kDa; TA-8; Targ4; transforming growth factor alpha regulated gene 4; tst; wu:fb99f09 | |
Rabbit | |
sequential chromatography | |
RUO | |
23451, 81898, 84486 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75533, Q99NB9 | |
SF3B1 | |
A synthesized peptide derived from human SF3B1(Accession O75533), corresponding to amino acid residues around phosphorylated Thr313. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |