missing translation for 'onlineSavingsMsg'
Learn More

Phosphocreatine disodium salt, Tocris Bioscience™

Product Code. 18004862
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Phosphate reservoir

Phosphate reservoir; used to regenerate ATP during skeletal muscle contraction.

Specifications

CAS 922-32-7
Molecular Formula C4H8N3Na2O5P
MDL Number MFCD00150192
Quantity 50 mg
Synonym creatine phosphate sodium salt
InChI Key RNTXMYSPASRLFT-UHFFFAOYSA-L
SMILES [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
IUPAC Name disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Molecular Weight (g/mol) 255.08
PubChem CID 131887655
Formula Weight 255.08
Percent Purity >98%
Chemical Name or Material Phosphocreatine disodium salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'