missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Phosphogluconic Acid
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
81.10 € - 554.00 €
Specifications
Chemical Name or Material | 6-Phosphogluconic Acid Trisodium Salt Dihydrate |
---|---|
CAS | 57775-17-4 |
Molecular Formula | C6H10Na3O10P-2H2O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018670
|
Melford
P20350-0.1 |
100 mg |
81.10 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018671
|
Melford
P20350-0.5 |
500 mg |
350.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018672
|
Melford
P20350-1.0 |
1 g |
554.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 53411-70-4
- Molecular Formula: C6H10O10PNa3 2H2O
- Molecular Weight: 378.11
Specifications
6-Phosphogluconic Acid Trisodium Salt Dihydrate | |
57775-17-4 | |
C6H10Na3O10P-2H2O |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title