missing translation for 'onlineSavingsMsg'
Learn More

Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals

Product Code. 10678824
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54723-94-3
Molecular Formula H80Mo12N3O41P-6
Molecular Weight (g/mol) 1960.994
MDL Number MFCD00150423
InChI Key BIQVMZMYTFVCJZ-UHFFFAOYSA-H
Synonym ammonium phosphomolybdate hydrate
PubChem CID 131664298
IUPAC Name azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate
SMILES N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo]

Specifications

Color Yellow
Packaging Plastic bottle
Quantity 1 kg
Linear Formula (NH4)3PMo12O40·xH2O
Physical Form Powder
Solubility Information Solubility in water: insoluble.
Formula Weight 1876.35
Chemical Name or Material Phosphomolybdic acid, ammonium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.