missing translation for 'onlineSavingsMsg'
Learn More
Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals Product Code.: 10572771

Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals

Product Code. 10572771
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10572771

Brand: Thermo Scientific Acros 174280250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 940-71-6
Molecular Formula Cl6N3P3
Molecular Weight (g/mol) 347.65
MDL Number MFCD00006474
InChI Key UBIJTWDKTYCPMQ-UHFFFAOYSA-N
Synonym phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer
PubChem CID 220225
IUPAC Name 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl

Specifications

Melting Point 111.0°C to 115.0°C
Quantity 25 g
Assay Percent Range 98%
Fieser 02,206; 04,386; 06,469
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ether, benzene, toluene
Formula Weight 347.65
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 1.98
Density 1.9800g/mL
Chemical Name or Material Phosphonitrilic chloride trimer
Show More Show Less
compliance-icons
Product Identifier
  • Phosphonitrilic chloride trimer
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.