missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 21.0°C |
Boiling Point | 158.0°C |
Density | 1.6800g/mL |
Heavy Metals (as Pb) | 10ppm max. |
Packaging | EcoSafPak™ Glass Bottle |
Reducing Substances | Passes Test |
Specific Gravity | 1.68 |
Quantity | 500 mL |
Linear Formula | H3PO4 |
Physical Form | Liquid |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467346 | Certificate of Analysis | 31/01/2025 | 424045000,424040100,424040025 |
A0463701 | Certificate of Analysis | 14/06/2024 | 424045000,424040100,424040025 |
B0541321A | Certificate of Analysis | 14/11/2023 | 424045000,424040100,424040025 |
B0541293A | Certificate of Analysis | 14/11/2023 | 424045000,424040100,424040025 |
A0457198 | Certificate of Analysis | 24/10/2023 | 424045000,424040100,424040025 |
Safety and Handling

Product Identifier
- Phosphoric acid, 85+% solution in water
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Orthophosphoric acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction