missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, Multi-Compendial, NF, J.T. Baker™

Product Code. 14801332
Click to view available options
Quantity:
500 mL
2.5 L
4 L
Packaging:
Glass Bottle
Glass Bottle, Single ship
PVC Coated Glass Bottle
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Packaging Glass Bottle, Single ship
Specific Gravity 1L = 1.69kg
Quantity 2.5 L
Formula Weight 98
Grade Multi-Compendial/NF
Chemical Name or Material Phosphoric Acid
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.