missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, N.F. - F.C.C., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Glass Bottle, Mall
Glass Bottle, Single ship
Unit Size:
2.5 L
500mL
Specifications
Specifications
| Packaging | Glass Bottle |
| Specific Gravity | 1L = 1.69kg |
| Quantity | 2.5 L |
| Formula Weight | 98 |
| Grade | FCC/NF |
| Chemical Name or Material | Phosphoric Acid |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction