missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss., meets analytical spec. of BP, FCC, NF, Ph. Eur., 85.0-88.0%, Honeywell Fluka™
BIOPHARMA

Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., 85.0-88.0%

113.94 € - 1131.71 €

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O
View More Specs

Products 8
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15640710
BIOPHARMA
View Documents
Honeywell-Fluka
04107-1L
1 L
113.94 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15650710
BIOPHARMA
missing translation for 'viewDocuments'
Honeywell-Fluka
04107-2.5L
2.5 L
288.35 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15690710
BIOPHARMA
missing translation for 'viewDocuments'
Honeywell-Fluka
04107-5L
5 L
462.45 €
5 L
Please to purchase this item. Need a web account? Register with us today!
15600720
missing translation for 'viewDocuments'
Honeywell-Fluka
04107-6X1L
6 x 1 L
541.69 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15670710
BIOPHARMA
missing translation for 'viewDocuments'
Honeywell-Fluka
04107-4X2.5L
4 x 2.5 L
854.91 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15630710
missing translation for 'viewDocuments'
Honeywell-Fluka
04107-10L
10 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15680710
missing translation for 'viewDocuments'
Honeywell-Fluka
04107-4X5L
4 x 5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15660710
BIOPHARMA
missing translation for 'viewDocuments'
Honeywell-Fluka
04107-45KG
45 kg
491.22 €
45kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7664-38-2
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
phosphoric acid
H3O4P
MFCD00011340
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
CHEBI:26078
OP(=O)(O)O

Specifications

7664-38-2
1.685g/mL at 25°C
H3O4P
MFCD00011340
1921286
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
1004
98g/mol
Phosphoric Acid, Meets analytical specification of Ph. Eur.BP, NF, FCC
85 to 88.0%
5mmHg (25°C), 2.2mmHg (20°C)
H3PO4
1805
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
OP(=O)(O)O
97.994
CHEBI:26078
puriss.
Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338-P310

H290-H314

missing translation for 'einecsNumber' : 231-633-2

Product Identifier
  • Phosphoric acid

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Orthophosphoric acid

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.