missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T), Honeywell Fluka™
Puriss. p.a., crystallized,≥99.0% (T)
22.91 € - 176.68 €
Chemical Identifiers
CAS | 7664-38-2 |
---|---|
Molecular Formula | H3O4P |
Molecular Weight (g/mol) | 97.994 |
MDL Number | MFCD00011340 |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico |
PubChem CID | 1004 |
ChEBI | CHEBI:26078 |
IUPAC Name | phosphoric acid |
SMILES | OP(=O)(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15607160
|
Honeywell-Fluka
79622-100G |
100 g | Glass Bottle | N/A | |||||
15627160
|
Honeywell-Fluka
79622-500G |
500 g | Glass Bottle |
105.22 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15617160
|
Honeywell-Fluka
79622-1KG |
1 kg | Glass bottle |
176.68 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7664-38-2 | |
97.994 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
1004 | |
phosphoric acid |
H3O4P | |
MFCD00011340 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
CHEBI:26078 | |
OP(=O)(O)O |
Specifications
7664-38-2 | |
158°C (literature) | |
≤100.0000% | |
2.2 mmHg (20°C), 5 mmHg (25°C) | |
H3PO4 | |
UN3453 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
≥99% | |
Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T) |
∼40°C (literature) | |
≥90.0000% | |
1.685 g/mL at 25 °C(lit.) | |
H3O4P | |
MFCD00011340 | |
1921286 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
98 | |
puriss. p.a. |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
missing translation for 'shelfLife' : 1620 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Phosphoric acid, solid
missing translation for 'einecsNumber' : 231-633-2
missing translation for 'rtecsNumber' : 231-633-2
Recommended Storage : Room Temp