missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, TraceSELECT™, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Specifications
Concentration | 85% |
Density | 1.685g/mL at 25°C |
Vapor Pressure | 5 mmHg (25°C), 2.2 mmHg (20°C) |
Quantity | 500 mL |
Linear Formula | H3PO4 |
UN Number | UN1805 |
Beilstein | 1921286 |
Formula Weight | 98g/mol |
Grade | TraceSELECT™ |
Chemical Name or Material | Phosphoric acid |
Safety and Handling

Product Identifier
- Phosphoric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Orthophosphoric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phosphoric acid, TraceSELECT™, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction