Learn More
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
69.24 € - 203.01 €
Chemical Identifiers
CAS | 13598-36-2 |
---|---|
Molecular Formula | H3O3P |
MDL Number | MFCD00137258 |
Synonym | Orthophosphorous acid |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10783201
|
Thermo Scientific Acros
201155000 |
500 g | Glass bottle |
69.24 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10496193
|
Thermo Scientific Acros
201150010 |
1 kg | Glass bottle |
126.63 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10072760
|
Thermo Scientific Acros
201150020 |
2 kg | Glass bottle |
203.01 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13598-36-2 | |
H3O3P | |
MFCD00137258 | |
Orthophosphorous acid |
Specifications
White | |
73.0°C | |
1.6500g/mL | |
1.65 | |
98% | |
H3PO3 | |
MFCD00137258 | |
Orthophosphorous acid | |
82 | |
Extra Pure |
13598-36-2 | |
200.0°C | |
20ppm max. | |
>100°C | |
H3O3P | |
Crystals | |
15, 7458 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
98% | |
Phosphorous acid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Phosphorous acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.