missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Pentasulfide, 99%, Honeywell™ Fluka™

Product Code. 15692300
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 1314-80-3
Molecular Formula P4S10
Molecular Weight (g/mol) 444.495
MDL Number MFCD00011441
InChI Key CYQAYERJWZKYML-UHFFFAOYSA-N
Synonym phosphorus pentasulfide, sulfur phosphide, diphosphorus pentasulfide, diphosphorus pentasulphide, tetraphosphorus decasulfide, phosphoric sulfide, phosphorus persulfide, phosphorus pentasulphide, thiophosphoric anhydride, sirnik fosforecny czech
PubChem CID 14817
SMILES P12(=S)SP3(=S)SP(=S)(S1)SP(=S)(S2)S3

Specifications

Boiling Point 515°C
Melting Point 280°C to 284°C
Color Green-Yellow to Yellow
Quantity 100 g
UN Number UN1340
Formula Weight 222.27g/mol
Percent Purity 99%
Packaging Glass Bottle
Appearance Powder
Density 2.09g/mL at 25°C
Chemical Name or Material Phosphorus Pentasulfide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'