missing translation for 'onlineSavingsMsg'
Learn More
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals Product Code.: 10124360

Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals

Product Code. 10124360
1 kg, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10124360

Brand: Thermo Scientific Acros 200890010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
Synonym Phosphoric anhydride
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Melting Point 340.0°C to 360.0°C
Color White
Physical Form Hygroscopic Crystalline Powder
Quantity 1 kg
Assay Percent Range 98%
Linear Formula P2O5
Fieser 01,871; 07,291; 10,319; 14,151; 15,267
Merck Index 15,7467
Solubility Information Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat.
Formula Weight 141.94
Percent Purity 98%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Phosphorus pentoxide
Show More Show Less
compliance-icons
Product Identifier
  • Phosphorus pentoxide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.