missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus tribromide, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

88.14 € - 322.61 €

Specifications

Chemical Name or Material Phosphorus tribromide
Name Note 1.0M Solution in Dichloromethane
CAS 7789-60-8, 75-09-2
Color Colorless to Yellow
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10108072
View Documents
Thermo Scientific Acros
382061000
100 mL AcroSeal™ Glass bottle
88.14 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10705993
View Documents
Thermo Scientific Acros
382068000
800 mL AcroSeal™ Glass Bottle
322.61 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

Phosphorus tribromide
7789-60-8, 75-09-2
Liquid
PBr2
15, 7469
IPNPIHIZVLFAFP-UHFFFAOYSA-N
tribromophosphane
24614
0.9 to 1.1M
1.4880g/mL
1.0M Solution in Dichloromethane
Colorless to Yellow
Br3P
MFCD00011436
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
P(Br)(Br)Br
270.69
270.69
1.488
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 232-178-2

Product Identifier
  • Phosphorus tribromide, 1M solution in dichloromethane

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only