missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus tribromide, 99%, Thermo Scientific Chemicals

Product Code. 10092180
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-60-8
Molecular Formula Br3P
Molecular Weight (g/mol) 270.69
MDL Number MFCD00011436
InChI Key IPNPIHIZVLFAFP-UHFFFAOYSA-N
Synonym phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
PubChem CID 24614
IUPAC Name tribromophosphane
SMILES P(Br)(Br)Br

Specifications

Boiling Point 175.0°C
Melting Point -40.0°C
Color Brown or Colorless to Yellow
Physical Form Liquid
Quantity 5 g
Assay Percent Range 98.5% min. (ex Br) (Argentometry)
Linear Formula PBr3
Fieser 01,873; 02,330; 07,292
Merck Index 15, 7469
Solubility Information Solubility in water: reacts. Other solubilities: soluble in acetone, alcohol, carbon disulfide,, hydrogen sulfide, methanol, ether, chloroform and, carbon tetrachloride
Formula Weight 270.69
Percent Purity 99%
Packaging Glass bottle
Refractive Index 1.697
Specific Gravity 2.88
Density 2.8800g/mL
Chemical Name or Material Phosphorus tribromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phosphorus tribromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'