missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Tribromide, Extra Pure, SLR, Fisher Chemical

Product Code. 10488410
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Coated Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Boiling Point 175°C
Melting Point -40°C
CAS 7789-60-8
Color Yellow
Physical Form Liquid
Quantity 100 mL
Molecular Formula Br3P
MDL Number 11436
Synonym phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
InChI Key IPNPIHIZVLFAFP-UHFFFAOYSA-N
SMILES P(Br)(Br)Br
IUPAC Name tribromophosphane
Molecular Weight (g/mol) 270.686
PubChem CID 24614
Formula Weight 270.69g/mol
Packaging Plastic Coated Glass bottle
Vapor Pressure 10mmHg 47.8
Density Vapor Density: 9.3
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phosphorus tribromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'