Learn More
Phosphorus(V) bromide, 95%, Thermo Scientific Chemicals
84.22 € - 906.51 €
Chemical Identifiers
CAS | 7789-69-7 |
---|---|
Molecular Formula | Br5P |
Molecular Weight (g/mol) | 430.494 |
MDL Number | MFCD00011437 |
InChI Key | QRKVRHZNLKTPGF-UHFFFAOYSA-N |
Synonym | phosphorus pentabromide, phosphorane, pentabromo, unii-3d9wis0bqw, pentabromophosphorane, phosphorus v bromide, 3d9wis0bqw, pentabromo, phosphorpentabromid, phosphoruspentabromide, phosphorouspentabromide |
PubChem CID | 62678 |
IUPAC Name | pentabromo-$l^{5}-phosphane |
SMILES | P(Br)(Br)(Br)(Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400667
|
Thermo Scientific Alfa Aesar
B22481.14 |
25 g |
84.22 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410667
|
Thermo Scientific Alfa Aesar
B22481.22 |
100 g |
215.07 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420667
|
Thermo Scientific Alfa Aesar
B22481.36 |
500 g |
906.51 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
7789-69-7 | |
430.494 | |
QRKVRHZNLKTPGF-UHFFFAOYSA-N | |
62678 | |
P(Br)(Br)(Br)(Br)Br |
Br5P | |
MFCD00011437 | |
phosphorus pentabromide, phosphorane, pentabromo, unii-3d9wis0bqw, pentabromophosphorane, phosphorus v bromide, 3d9wis0bqw, pentabromo, phosphorpentabromid, phosphoruspentabromide, phosphorouspentabromide | |
pentabromo-$l^{5}-phosphane |
Specifications
105°C to 107°C (decomposition) | |
7789-69-7 | |
MFCD00011437 | |
3903332 | |
phosphorus pentabromide, phosphorane, pentabromo, unii-3d9wis0bqw, pentabromophosphorane, phosphorus v bromide, 3d9wis0bqw, pentabromo, phosphorpentabromid, phosphoruspentabromide, phosphorouspentabromide | |
QRKVRHZNLKTPGF-UHFFFAOYSA-N | |
pentabromo-$l^{5}-phosphane | |
62678 | |
95% | |
Moisture sensitive |
∽105°C (decomposition) | |
Br5P | |
UN2691 | |
14,7350 | |
Reacts with water. | |
P(Br)(Br)(Br)(Br)Br | |
430.494 | |
430.5 | |
Acrid | |
Phosphorus(V) bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS PENTABROMIDE
missing translation for 'einecsNumber' : 232-186-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Phosphorus(V) bromide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : Phosphorus pentabromide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.