Learn More
Phosphorus(V) oxide, ACS, 98% min, Thermo Scientific Chemicals
For Use As a Drying and Dehydrating Agent, As a Condensing Agent in organic Synthesis, in Sugar Refining
Chemical Identifiers
CAS | 1314-56-3 |
---|---|
Molecular Formula | O5P2 |
Molecular Weight (g/mol) | 141.94 |
MDL Number | MFCD00011440 |
InChI Key | DLYUQMMRRRQYAE-UHFFFAOYSA-N |
IUPAC Name | tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone |
SMILES | O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366038
|
Thermo Scientific Alfa Aesar
010524.18 |
50 g |
N/A
|
N/A | |||||
11386038
|
Thermo Scientific Alfa Aesar
010524.36 |
500 g |
N/A
|
N/A | |||||
11376038
|
Thermo Scientific Alfa Aesar
010524.57 |
4 x 500 g |
N/A
|
N/A | |||||
Description
Used as a drying and dehydrating agent, condensation reagent in organic synthesis and a laboratory reagent. Used in sugar refining, and in fire extinguishing. Phosphorus pentoxide in DMSO forms Onodera reagent, which oxidizes alcohols. Capable of converting mineral acids to anhydrides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-56-3 | |
141.94 | |
DLYUQMMRRRQYAE-UHFFFAOYSA-N | |
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3 |
O5P2 | |
MFCD00011440 | |
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone |
Specifications
1314-56-3 | |
Powder | |
O5P2 | |
Phosphoric anhydride, Phosphoric oxide | |
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3 | |
141.94 | |
ACS | |
White powder |
White | |
98% min. | |
MFCD00011440 | |
DLYUQMMRRRQYAE-UHFFFAOYSA-N | |
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone | |
141.94 | |
Odorless | |
Phosphorus(V) oxide |
Safety and Handling
H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 215-236-1
RUO – Research Use Only