missing translation for 'onlineSavingsMsg'
Learn More

Phosphotungstic Acid, 44-hydrate, Thermo Scientific™

Product Code. 10220041
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color White to Yellow
CAS 12067-99-1
Heavy Metals (as Pb) 50ppm max.
Packaging Glass bottle
Quantity 25 g
Molecular Formula 24WO3·44H2O
Linear Formula 24WO3·44H2O
Physical Form Crystalline Powder
MDL Number MFCD00149916
Merck Index 15, 7476
Synonym PTA
Solubility Information Solubility in water: practical insoluble: 0.02% max.
Formula Weight 6498.96
Chemical Name or Material Phosphotungstic acid, 44-hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Phosphotungstic acid, 44-hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.