Learn More
Cole-Parmer™ FF-200D-I Economical Flame Photometer
The FF-200D-I Economical Flame Photometer with automatic gas shutoff feature ensures operator safety.
Brand:  Cole-Parmer™ 83055-05
Product Code. 11485059
  • 5625.00 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unit comes with Na+, K+, Li+, Ca+, and Ba+ filters.
  • Low temperature and single channel.
  • Use this economical flame photometer for routine determinations of sodium and potassium in industrial and educational applications.
  • The fine and coarse adjustment controls provide accurate measurements every time.
  • For safety, an automatic gas shutoff mechanism activates if the flame becomes accidentally extinguished.
Specifications
Specifications
230 VAC
50/60 Hz
Photometer Flame of Industry
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ FF-200D-I Economical Flame Photometer > FF-200D-I Economical Flame Photometer; Electrical Requirements: 230 VAC, 50/60 Hz