missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ pHrodo™ Green Maleimide

pHrodo™ Green Maleimide

Brand:  Molecular Probes™ P35370

443.52 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 13345654

  • 672.00 € / 1mg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The thiol-reactive pH-sensitive pHrodo™ Green Maleimide dye is suitable for the creation of bioconjugates to study endocytosis and phagocytosis. pHrodo™ Green dramatically increases fluorescence as the pH of its surroundings become more acidic.
  • Use pH-sensitive pHrodo™ Green Maleimide to make pH-sensitive bioconjugates of your choice
  • Get faster, more accurate results than with any other endocytosis or phagocytosis assay—no need for wash steps or quenchers
  • Multiplex with red fluorescent markers such as RFP, pHrodo™ Red, and many others

    The increase in fluorescence of pHrodo™ Green as pH changes from basic to acidic correlates with the acidification of intracellular vesicles, making it an ideal tool to study endocytosis or phagocytosis and their regulation by environmental factors, drugs, or pathogens. The spectral properties of pHrodo™ Green makes it useful for multi-color experiments. pHrodo™ Green can be detected using most standard green fluorescent filter sets and has been validated for use on a variety of platforms including flow cytometry, fluorescent microscopy, and high content screening (HCS). The lack of fluorescence of pHrodo™ Green in a typical extracellular environment eliminates the need for wash steps or quencher dyes in the experimental workflow.

    pHrodo™ Green Maleimide is a thiol-reactive dye that can be used to create pHrodo™ Green bioconjugates in aqueous buffer. The maleimide reacts with free sulphydryl groups produced by the reduction of cysteines in proteins or peptides. Maleimides are particularly useful for labeling antibodies as the dye will not attach to the antibody binding site. This reaction will result in a stable conjugate that can be used in live cell assays or stored for later use.

    pHrodo™ Green is also available in an amine-reactive form (see pHrodo™ Green STP), as well as a selection of ready-to-use conjugates (e.g., E. coli, S. aureus, and dextran). In addition, pHrodo™ Red reactive dyes and ready-to-use conjugates are available as a color alternative with the same properties.

    For Research Use Only. Not for human or animal therapeutic or diagnostic use.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Dried Down
    Maleimide
    1 vial , store ≤ -20 °C, dessicate and protected from light.
    Fluorescence
    Thiol
    Green
    525
    1 mg
    DMSO (Dimethylsulfoxide)
    Thiol
    75000
    pHrodo™ Green
    pHrodo™
    505
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.