missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ pHrodo™ Red Maleimide
pHrodo™ Red Maleimide
Brand: Molecular Probes™ P35371
434.28 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The thiol-reactive pH-sensitive pHrodo™ Red Maleimide dye is suitable for the creation of bioconjugates to study endocytosis and phagocytosis. pHrodo™ Red dramatically increases fluorescence as the pH of its surroundings become more acidic.The increase in fluorescence of pHrodo™ Red as pH changes from basic to acidic correlates with the acidification of intracellular vesicles, making it an ideal tool to study endocytosis or phagocytosis and their regulation by environmental factors, drugs, or pathogens. The spectral properties of pHrodo™ Red makes it useful for multi-color experiments. pHrodo™ Red has been validated for use on a variety of platforms including flow cytometry, fluorescent microscopy, and high content screening (HCS). The lack of fluorescence of pHrodo™ Red in a typical extracellular environment eliminates the need for wash steps or quencher dyes in the experimental workflow.
pHrodo™ Red Maleimide is a thiol-reactive dye that can be used to create pHrodo™ Red bioconjugates in aqueous buffer. The maleimide reacts with free sulphydryl groups produced by the reduction of cysteines in proteins or peptides. Maleimides are particularly useful for labeling antibodies as the dye will not attach to the antibody binding site. This reaction will result in a stable conjugate that can be used in live cell assays or stored for later use.
pHrodo™ Red is also available in an amine-reactive form (see pHrodo™ Red SE), as well as a selection of ready-to-use conjugates (e.g., E. coli, S. aureus, and dextran). In addition, pHrodo™ Green reactive dyes and ready-to-use conjugates are available as a color alternative with the same properties.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Dried Down | |
Maleimide | |
1 vial , store ≤ -20 °C, dessicate and protected from light. | |
Fluorescence | |
Thiol | |
Red | |
585 |
1 mg | |
DMSO (Dimethylsulfoxide) | |
Thiol | |
65000 | |
pHrodo™ Red | |
pHrodo™ | |
560 |
For Research Use Only. Not for use in diagnostic procedures.