missing translation for 'onlineSavingsMsg'
Learn More
Phthalan, 98%, Thermo Scientific Chemicals Product Code.: 11439552

Phthalan, 98%, Thermo Scientific Chemicals

Product Code. 11439552
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11439552

Brand: Thermo Scientific Alfa Aesar A10217.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4'-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4′-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

Solubility
Low aqueous solubility.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-14-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005932
InChI Key SFLGSKRGOWRGBR-UHFFFAOYSA-N
Synonym phthalan, 1,3-dihydroisobenzofuran, isocoumaran, isobenzofuran, 1,3-dihydro, o-xylylene oxide, 2-oxaindan, 1,3-dihydro isobenzofuran, unii-2r6ntq7y6g, ccris 1557, 2r6ntq7y6gShow MoreShow Less
PubChem CID 10327
IUPAC Name 1,3-dihydro-2-benzofuran
SMILES C1C2=CC=CC=C2CO1

Specifications

Density 1.094
Boiling Point 192°C
Flash Point 63°C (145°F)
Refractive Index 1.546
Quantity 100 g
Beilstein 111932
Solubility Information Low aqueous solubility.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material Phthalan

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10119504 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10217.22,A10217.14,A10217.06
Lot Number10111391 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10217.22,A10217.14,A10217.06
Lot NumberI6038A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10217.22,A10217.14,A10217.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phthalan, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.