missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalates in Polyethylene Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CRM-PE001
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Diisodecyl phthalate | 26761-40-0 | 30,000μg/g |
Diisononyl phthalate | 28553-12-0 | 30,000μg/g |
Bis(2-Ethylhexyl)phthalate | 117-81-7 | 3000μg/g |
Butylbenzyl phthalate | 85-68-7 | 3000μg/g |
Diethyl phthalate | 84-66-2 | 3000μg/g |
Dimethyl phthalate | 131-11-3 | 3000μg/g |
Di-n-butyl phthalate | 84-74-2 | 3000μg/g |
Di-n-octyl phthalate | 117-84-0 | 3000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Phthalates in Polyethylene Standard | |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
Polyethylene |
Designed to be used with the current regulations and testing methods including: US Method: CPSC-CH-C1001.09.3 EU Directive: 2005/84/EC |
|
5 g | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- phthalates in polyethylene standard
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1
Hazard Statement
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : di-n-octyl phthalate, BBP, dibutyl phthalate, bis(2-ethylhexyl) phthalate
Product intended for laboratory use only