missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, 99%, Thermo Scientific Chemicals

35.10 € - 301.00 €

Chemical Identifiers

CAS 88-99-3
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002467
InChI Key XNGIFLGASWRNHJ-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
PubChem CID 1017
ChEBI CHEBI:29069
IUPAC Name phthalic acid
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11487107
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14450.30
250 g
35.10 €
250g
Please to purchase this item. Need a web account? Register with us today!
11477107
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14450.0B
1000 g
91.60 €
1000g
Please to purchase this item. Need a web account? Register with us today!
11497107
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14450.0I
5000 g
301.00 €
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

Solubility
Soluble in water, alcohol and etherSoluble in alcohol. Slightly soluble in water, methanol and ether. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

88-99-3
166.132
XNGIFLGASWRNHJ-UHFFFAOYSA-N
1017
phthalic acid
C8H6O4
MFCD00002467
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
CHEBI:29069
C1=CC=C(C(=C1)C(=O)O)C(=O)O

Specifications

88-99-3
C8H6O4
608199
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
XNGIFLGASWRNHJ-UHFFFAOYSA-N
phthalic acid
1017
166.13
1.593
Odorless
205°C (decomposition)
MFCD00002467
14,7371
Soluble in water,alcohol and etherSoluble in alcohol; Slightly soluble in water,methanol and ether; Insoluble in chloroform.
C1=CC=C(C(=C1)C(=O)O)C(=O)O
166.132
CHEBI:29069
99%
168°C (334°F)
Phthalic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

missing translation for 'einecsNumber' : 201-873-2

missing translation for 'rtecsNumber' : TH9625000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only