missing translation for 'onlineSavingsMsg'
Learn More

Phthalic anhydride, 99%, Thermo Scientific™

Product Code. 11407157
Click to view available options
250 g

 

Phthalic anhydride is used in the preparation of primary amines. It is used as a precursor to prepare anthraquinone, phthalein, rhodamine, phthalocyanine, fluorescein and xanthene dyes. It serves as an important intermediate in the plastics industry, dyes, resins and pigments. It is used as a monomer for synthetic resins such as glyptal, alkyd resins and polyester resins. It plays a vital role in the large-scale production of plasticizers for plastics. Further, it acts as a curing agent for epoxy resins, anti-scaling agents, plating agents, surface treating agents, ion exchange agents, paint additives, adhesives, sealant chemicals and potential antibacterial agent.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Phthalic anhydride is used in the preparation of primary amines. It is used as a precursor to prepare anthraquinone, phthalein, rhodamine, phthalocyanine, fluorescein and xanthene dyes. It serves as an important intermediate in the plastics industry, dyes, resins and pigments. It is used as a monomer for synthetic resins such as glyptal, alkyd resins and polyester resins. It plays a vital role in the large-scale production of plasticizers for plastics. Further, it acts as a curing agent for epoxy resins, anti-scaling agents, plating agents, surface treating agents, ion exchange agents, paint additives, adhesives, sealant chemicals and potential antibacterial agent.

Solubility
Soluble in water, carbon disulfide, ethanol, acetone and benzene. Slightly soluble in ethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
TRUSTED_SUSTAINABILITY

Specifications

CAS 85-44-9
Melting Point 130°C to 134°C
Density 1.53
Boiling Point 295°C (sublimation)
Flash Point 152°C
Odor Weak, Characteristic
Molecular Formula C8H4O3
MDL Number MFCD00005918
Quantity 250 g
UN Number UN2214
Beilstein 118515
Sensitivity Moisture sensitive
Merck Index 14,7372
Synonym phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione
Solubility Information Soluble aq. soln.,carbon disulfide,ethanol,acetone and benzene. Slightly soluble in ethyl ether.
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
SMILES O=C1OC(=O)C2=CC=CC=C12
IUPAC Name 1,3-dihydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 148.12
PubChem CID 6811
ChEBI CHEBI:36605
Formula Weight 148.12
Percent Purity 99%
Chemical Name or Material Phthalic anhydride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phthalic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Phthalic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.