missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic anhydride, ACS, 99.0-100.2%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
4 x 500 g
Description
Phthalic anhydride is used as an intermediate in chemical synthesis and plastics manufacturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 131°C to 133°C |
Density | 1.53 g/mL |
Boiling Point | 295°C (sublimation) |
Flash Point | 152°C (305°F) |
Odor | Weak, characteristic |
Quantity | 500 g |
UN Number | UN2214 |
Beilstein | 118515 |
Sensitivity | Moisture sensitive |
Merck Index | 14,7372 |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction