missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion™ Hot Start II DNA Polymerases (2 U/μL)

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

152.00 € - 2305.00 €

Specifications

GC-Rich PCR Performance High
Reaction Speed Fast
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Concentration 2 U/μL
View More Specs

Includes: Phusion Hot Start II High–Fidelity DNA Polymerase
  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/μL)
  • 5x Phusion HF Buffer
  • 5x Phusion GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Phusion Green Hot Start II High–Fidelity DNA Polymerase
  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/Glc muL)
  • 5x Phusion Green HF Buffer
  • 5x Phusion Green GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Both Phusion and Phusion Green Buffer provide 1.5 mM MgCl2 in the final 1x concentration

Products 5
Product Code Brand Color No. of Reactions Price Quantity & Availability  
Product Code Brand Color No. of Reactions Price Quantity & Availability  
13505170
View Documents
Thermo Scientific™
F537S
Green 100 Reactions
156.00 €
100 units
Please to purchase this item. Need a web account? Register with us today!
13595160
View Documents
Thermo Scientific™
F537L
Green 500 Reactions
688.00 €
500 units
Please to purchase this item. Need a web account? Register with us today!
10441338
View Documents
Thermo Scientific™
F549S
Colorless 100 Reactions
152.00 €
100 units
Please to purchase this item. Need a web account? Register with us today!
10628439
View Documents
Thermo Scientific™
F549L
Colorless 500 Reactions
688.00 €
500 units
Please to purchase this item. Need a web account? Register with us today!
16258310
View Documents
Thermo Scientific™
F549XL
Colorless 2000 Reactions
2305.00 €
2000 units
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

High
Dry Ice
2 U/μL
Built-In Hot Start
Standalone
Fast
Hot-start PCR, High-fidelity PCR
52X
Blunt
20 kb or less
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Phusion™ Hot Start II DNA Polymerases (2 U/μL)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.