missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion High-Fidelity PCR Kit

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

Brand:  Thermo Scientific™ F553L

198.25 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10084207

  • 305.00 € / 200 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Phusion High-Fidelity DNA Polymerase sets a high standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is an excellent choice for cloning and other applications requiring high fidelity. Phusion DNA polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerases.

The Phusion High-Fidelity PCR Kit contains all the necessary reagents for PCR including control lambda DNA template and primers for 1.3 kb and 10 kb amplicons. The DNA template amount is sufficient for performing 20 control reactions in 50 μL volume or 50 control reactions in 20 μL volume.

Highlights

  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15–30 s/kb)
  • Robust performance, minimal optimization needed
  • Increased product yields with minimal enzyme amounts

Applications

  • High-fidelity PCR
  • High throughput
  • Amplification of difficult (GC-rich) templates
  • Template generation for sequencing
  • Multiplex PCR
  • Long-range PCR
  • Cloning & mutagenesis
  • Microarray

Notes

  • Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Phusion DNA Polymerase (2 U/μL)
• 5X Phusion GC & HF Buffer
• dNTP mix (10 mM each)
• Control template (λ DNA) and DNA size standard
• 1.3 kb & 10 kb primers (4 μM each)
• 50 mM MgCl2 solution and DMSO

Store at –20°C.
Phusion High-Fidelity DNA Polymerase
High-Fidelity PCR Kit
Standard PCR, High-fidelity PCR
No
Blunt
20 kb or less
High
Fast
Dry Ice
52X
200 Reactions
Separate Components
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.