missing translation for 'onlineSavingsMsg'
Learn More
uvex Phynomic C3 Cut Protection Gloves Product Code.: 17270381
Estimated Shipment: 30-05-2025
to see stock.
Add to basket

uvex Phynomic C3 Cut Protection Gloves

Product Code. 17270381 Shop All Uvex Products
6
Click to view available options
Size:
6
7
8
9
10
11
12
This item is not returnable. View return policy

Product Code. 17270381

Brand: uvex 6008006

Estimated Shipment: 30-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight, sensitive gloves provide level 3/B cut resistance for mechanical activities. Aqua-polymer foam coating is breathable and enhances grip capabilities in dry or slightly damp applications.

  • Lightweight construction acts as a second-skin for tactile sensitivity
  • Level 3/B cut protection for mechanical tasks
  • Aqua-polymer foam coating is breathable and repels dampness
  • Gloves are made from materials that prevent excessive perspiration
  • Enhanced grip in dry, and slightly damp applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Light Blue, Gray
Hand Ambidextrous
Latex Free Yes
Material Polyamide, Elastane, HPPE
Product Type Cut-resistant Gloves
Size 6
EN388 Standard 4X43B
For Use With (Application) Machine Operation, Ground Covering/Tilling
Gender Unisex
Series Phynomic C3
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
uvex Phynomic C3 Cut Protection Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.