missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Phynomic Pro Safety Gloves

Product Code. 17230381
Click to view available options
Size:
6
7
8
9
10
11
12
This item is not returnable. View return policy
This item is not returnable. View return policy

Second skin design is light and flexible to help minimize excessive perspiration, while aqua-polymer foam coating repels dirt and moisture.

  • Lightweight construction acts as a second-skin for tactile sensitivity
  • Bamboo/polyamide/elastane liner provides a high level of user comfort
  • Natural fibers help to absorb water vapor
  • Aqua-polymer foam coating repels dirt and moisture
  • Breathable materials help prevent excessive perspiration
  • Enhanced grip in dry, damp, and oily applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Pro
Color Blue, Gray
Material Bamboo, Polyamide, Elastane
Size 10
EN388 Standard 2121X
For Use With (Application) Assembly, Construction, Maintenance, Outdoor Activities, and Repairs
Gender Unisex
Glove Cuff Style Knit
Series Phynomic Pro
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'