missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic WET Water-Resistant Gloves with Aqua-Polymer Foam Coating

Product Code. 17270371
Click to view available options
Size:
10
11
12
This item is not returnable. View return policy
This item is not returnable. View return policy

Uvex™ Phynomic series gloves are ideal for all types of work where a high level of feeling is required. These safety gloves act as a second skin and are especially light and flexible, without making hands immediately sweaty. The aqua-polymer foam coating offers a high level of protection against humidity.

  • Water-repellent safety gloves with an aqua-polymer foam coating for use in outdoor areas
  • Breathable coating is durable to provide a high degree of mechanical abrasion resistance
  • Enhanced grip capabilities in damp and wet areas
  • Tactile feel when assembling parts
  • Ideal for precision work, maintenance, and repairs
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Blue, Black
Material Polyamide, Elastane
Size 12
EN388 Standard 3131X
For Use With (Application) Assembly, Maintenance, Precision Work, and Repairs
Gender Unisex
Glove Cuff Style Knit
Series Phynomic Wet
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.