Learn More
MP Biomedicals™ Phytohemagglutinin, Phaseolus Vulgaris
Phytohemagglutinin, Phaseolus Vulgaris is crude Phaseolus vulgaris-P lectin (PHA-P) from the Red Kidney Bean.
Brand:  MP Biomedicals™ 08791501
Product Code. 11464196
  • 781.00 € / 25mg
Explore more special offers
Description
Description
  • Crude Phaseolus vulgaris-P lectin (PHA-P) from the Red Kidney Bean.
  • Phytohemagglutinin PHA agglutinates the erythrocytes of all human blood groups as well as those of rabbit, dog, cat, mouse, sheep, and guinea pig.
  • The optimal amount of PHA required for the agglutination of erythrocytes varies with the blood from different individuals.
  • Protein or Enzyme Type: Other Proteins, Enzymes Peptides
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mg
Lyophilized
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.