Learn More
Buerkle™ Pi-Pump™
Proven thousands of times, Buerkle™ Pi-Pump™ fits naturally into the hand.
Brand: Buerkle™ 7551-0025
This item is not returnable.
View return policy
Description
- Handwheel ensures precise suction
- Contents can be purged by pressing back the piston
- Dispensing with the lateral switch
- Green
- Easy to use
Specifications
Red | |
Pipette | |
Pipetting up to 25 mL |