missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Piceatannol
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
Beschreibung
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
CAS | 10083-24-6 |
Melting Point | 235.0°C |
Molecular Formula | C14H12O4 |
Quantity | 250 mg |
Solubility Information | Solubility aq. soln.: insoluble. Other solubilities: soluble in ethanol, dmso, dimethyl formamide,, acetone, ethyl acetate, benzene |
InChI Key | CDRPUGZCRXZLFL-OWOJBTEDSA-N |
SMILES | OC1=CC(\C=C\C2=CC=C(O)C(O)=C2)=CC(O)=C1 |
IUPAC Name | 5-[(1E)-2-(3,4-dihydroxyphenyl)ethenyl]benzene-1,3-diol |
Molecular Weight (g/mol) | 244.25 |
Formula Weight | 244.24 |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'