missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PichiaPink™ Secretion Signal Set

The PichiaPink™ Secretion Signal Set is a set of 8 DNA fragments encoding secretion signal sequences. The PichiaPink™ Secretion Signal Set is designed to be easily ligated in front of your gene.

Brand:  Invitrogen™ A11155

Product Code. 10465785

  • 2565.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The PichiaPink™ Secretion Signal Set is a set of 8 DNA fragments encoding secretion signal sequences. The PichiaPink™ Secretion Signal Set is designed to be easily ligated in front of your gene. The PichiaPink™ Secretion Signal Set is designed for use with the pPINK-HC and pPINK-LC vectors.

Optimize Secretion of Your Expressed Protein

By adding one of the 8 secretion signal sequences to your gene, you can secrete your protein into the media to make it easier to harvest and purify. Pichia pastoris secretes few native proteins, so the bulk of the protein in the media will be yours.

Each DNA sequence encodes a different secretion signal. You can test each secretion signal sequence to find the one that works best for your protein.

The PichiaPink™ Secretion Signal Set comes with 8 different secretion signals including:

  • α-mating factor pre-sequence from Saccharomyces cerevisiae
  • α-amylase signal sequence from Aspergillus niger
  • Glucoamylase signal sequence from Aspergillus awamori
  • Serum albumin signal sequence from Homo sapiens
  • Inulinase signal sequence from Kluyveromcyes maxianus
  • Invertase signal sequence from Saccharomyces cerevisiae
  • Killer protein signal sequence from Saccharomyces cerevisiae
  • Lysozyme signal sequence from Gallus gallus

Easily Add a Secretion Signal to Your Protein

The DNA fragments in the PichiaPink™ Secretion Signal Set are designed to be easily inserted in front of your gene when using the pPINK-HC or pPINK-LC vectors. You can try different secretion signal sequences to see which one gives optimal secretion of your protein.

Each signal sequence includes the Kozak sequence for the AOX1 gene from Pichia pastoris to ensure efficient translation of your protein.

The secretion signal sequences come as phosphorylated duplex oligomers in 40 pmol aliquots. Resuspend oligomers in 40 μL TE buffer before use.

Why Choose the PichiaPink™ Yeast Expression System?

The PichiaPink™ Yeast Expression System is based on the yeast Pichia pastoris. Advantages of Pichia pastoris include rapid growth, well-defined genetic background, simple media formulation, and easy handling. For over 30 years, Pichia pastoris has been used by labs around the world for producing hundreds of different proteins from many species including human (Ref 1, 2). The PichiaPink™ Yeast Expression System allows convenient and cost-effective protein production from small to large scales.

For information on obtaining a commercial-use license for the PichiaPink™Yeast Expression System, please inquire at outlicensing@lifetech.com.

References

1. Cereghino JL, Cregg JM. Heterologous protein expression in the methylotrophic yeast Pichia pastoris. FEMS Microbiol Rev. 2000 Jan;24(1):45-66. [PubMed]

2. Cereghino GP, Cereghino JL, Ilgen C, Cregg JM. Production of recombinant proteins in fermenter cultures of the yeast Pichia pastoris. Curr Opin Biotechnol. 2002 Aug;13(4):329-32. [PubMed]

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PichiaPink Secretion Signal Set, 1 Kit, Designed to be Easily Ligated in Front of your Gene, with the pPINK-HC and pPINK-LC Vectors, Optimize Secretion of Your Expressed Protein, Set of 8 DNA Fragments Encoding Secretion Signal Sequences, Cloning into expression vectors
PichiaPink Secretion Signal Set, Secretion Signal Set
P. pastoris
The PichiaPink™ Secretion Signal Set consists of 8 signal sequences which are DNA duplexes:
• S. cerevisiae Alpha mating factor presequence
• Asp. niger alpha Amylase
• Asp. awarmori Glucoamylase signal
• H. sapien serum albumin signal
• K. maxianus Inulinase presequence
• S.cerevisiae Killer Protein sequence
• S.Cerevisiae invertase Signal sequence
• G. gallus Lysozyme Signal sequence

Quantity: 40 pMoles
Store at -20°C.
AOX1
Cell-Based Expression
Protein biology, protein expression, yeast expression, pichia pastoris expression systems, cloning into expression vectors, Protein Expression
1 Kit, 8 DNA duplexes kit
PichiaPink™
Restriction Enzyme/MCS
pPINK-LC, pPINK-HC
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.