missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Picking Bin

Product Code. 17988991
Click to view available options
Color:
Blue
Gray
Green
Red
White
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17988991

Brand: Gilac™ G152836

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Picking Bin is open fronted for easily accessible contents. 6 available colors.

  • Open fronted picking bins.
  • Contents easily accessible.
  • Stackable up to 2 metres.
  • Stability reinforced through bin feet interlocking.
  • Available in 6 colors, making it easier to differentiate the contents.

Specifications

Type Storage Bin
Material HDPE
Color Green
Shape Rectangular
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.