Learn More
Thermo Scientific™ Pico™ 17 Microcentrifuge
GREENER_CHOICE
The compact footprint of this microcentrifuge is ideal for routine laboratory work up to 17,000 xg.
Brand:  Thermo Scientific™ 75002414
1988.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 13496248
  • 2485.00 € / Each
Explore more special offers
Description
Description
Ideal for everyday laboratory use, Thermo Scientific Pico 17 microcentrifuges provide outstanding processing power, unique safety features and a broad array of microcentrifuge rotors and accessories for microcentrifuge tube processing.
Support your micro-volume separation needs with:
  • Performance up to 17,000 x g with fast acceleration and deceleration.
  • Choose from 7 optional microcentrifuge rotors to find the ideal fit for your application:
    • Outstanding corrosion resistance with lightweight engineered polymer rotors.
    • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns.
    • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids.
    • Eliminate the need for adapters with unique dual row rotor.
  • Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure.
  • Sleek, lightweight, space-saving design.
  • Durable and easy to clean and maintain.
  • Quiet performance promotes a safe, stress-free environment.
  • Conforms to the latest clinical and safety standards, including 98/79/EC In Vitro Diagnostic Directive.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 x 1.5/2 mL
17,000 x g
Ambient temperature
Benchtop Centrifuge
Pico 17 Centrifuge, Ventilated
No
IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
23.1lb. (10.5 kg)
50/60 Hz
Dual Row 18 x 2/0.5 mL Rotor with Screw-On Lid
13,300 rpm
56 dB(A)
CSA, CE, IVD
Dual Row 18 x 2.0/0.5mL rotor with screw-on lid
230 V 50/60 Hz
1 to 99 min. (1 min. increments); HOLD
8.9 x 9.6 x 13.8 in. (22.5 x 24.3 x 35.2 cm)
Pico
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products