Learn More
Applied Biosystems™ PicoPure™ RNA Isolation Kit
PicoPure™ RNA Isolation Kit
Brand:  Applied Biosystems™ KIT0204
 View more versions of this product
Product Code. 15295033
  • 452.00 € / 1 set
Explore more special offers
Description
Description
The ARCTURUS™ PicoPure™ RNA Isolation Kit is designed to recover high-quality total RNA consistently from fewer than ten cells, even from a single cell. This kit is designed to provide consistent, efficient RNA recovery without sacrificing quality. Small volume elution allows you to maximize your recovery of RNA from small numbers of cells for use with your gene expression analysis.
Key Product Features:
  • Greater sensitivty—isolate RNA from even a single cell
  • Superior efficiency—maximize RNA recovery from samples
  • Streamlined concentration—elute with small volumes
  • Flexible application—compatible with many tube sizes
  • Integrated contamination prevention—column lids ensure that samples remain RNase free
    Isolate RNA from Even a Single Cell
    The PicoPure™ RNA Isolation Kit is engineered to recover high-quality total RNA consistently from fewer than ten cells. The high recovery rate attained using this kit is valuable not only for small samples containing picograms of RNA but also for samples containing up to 100μg of RNA. Since the RNA elutes in as little as 10μl of buffer, it is ready for gene expression analysis without any vacuum concentration. This kit is available in two sizes, enough for 40 or 200 isolations.
    Get Superior RNA Recovery
    Small, valuable samples often cannot be replaced. Performing gene expression studies on these precious samples requires getting maximum RNA recovery. Comparative studies show that, with small, RNA-poor cell or tissue samples, the PicoPure™ Kit dramatically outperforms other commercial total RNA isolation kits in efficiency.
    Eliminate Time-consuming RNA Concentration
    The simple isolation protocol consists of extracting cellular RNA, then loading the extract onto the specially designed MiraCol™ Purification Column to bind the RNA. After washing away impurities, the RNA elutes in only 10μl of buffer, ready for use without vacuum concentration or transfer into a new reaction tube.
    High Yield Purification Columns
    The PicoPure™ RNA Isolation Kit features unique, high-recovery MiraCol™ Purification Columns, engineered to maximize recovery from microscopic samples. The columns have convenient integrated lids to help ensure that samples remain RNase free. In addition, the Kit comes complete with all necessary buffers, an easy-to-follow User Guide, optimized protocols for handling Laser Capture Microdissected (LCM) samples, and protocols for isolating RNA from larger samples.
    Deliver RNA Ready for Amplification and Microarray Analysis
    RNA isolated with the PicoPure™ RNA Kit is ready, without vacuum concentration, for amplification with the RiboAmp™ RNA Amplification Kit. The two kits together allow for highly reproducible gene expression results.
    Retain Low-abundance mRNA
    The PicoPure™ RNA Isolation Kit preserves mRNA in all abundance classes through the isolation process, minimizing the loss of important genes of interest. Low, medium, and high-abundance genes are consistently recovered and available for expression analysis.
    Maintain High RNA Quality
    The PicoPure™ RNA isolation process allows for high-quality RNA. Total cellular RNA prepared from LCM samples remains intact after isolation, increasing confidence in subsequent molecular analysis.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    Extraction buffer, purification reagents, purification columns, microcentrifuge tubes. Store at room temperature
    Spin Column
    Total RNA
    ArcturusXT™ LCM Instrument
    40 samples
    40 samples
    Total RNA
    Spin Column
    Laser Capture Microdissection (LCM), Tissue (Frozen)
    Not High-throughput Compatible (Manual)
    Microarray Analysis, Real-Time Quantitative PCR (qPCR)
    PicoPure™
    As little as a single cell.
    Product Suggestions
    Product Suggestions
    Videos
    Safety and Handling
    Safety and Handling
    Product Identifier
    • RNA Isolation Kit
    Signal Word
    • Danger
    Hazard Category
    • Acute toxicity Category 4
    • LONG-TERM AQUATIC HAZARD Chronic 3
    • Serious eye damage/eye irritation Category 2
    • Flammable liquid Category 2
    • Flammable liquid Category 3
    • Skin corrosion/irritation Category 1 C
    • Skin corrosion/irritation Category 2
    • Skin sensitiser Category 1
    Hazard Statement
    • H225-Highly flammable liquid and vapour.
    • H226-Flammable liquid and vapour.
    • H302-Harmful if swallowed.
    • H314-Causes severe skin burns and eye damage.
    • H315-Causes skin irritation.
    • H317-May cause an allergic skin reaction.
    • H319-Causes serious eye irritation.
    • H412-Harmful to aquatic life with long lasting effects.
    Precautionary Statement
    • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
    • P233-Keep container tightly closed.
    • P242-Use only non-sparking tools.
    • P243-Take precautionary measures against static discharge.
    • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
    • P264-Wash hands thoroughly after handling.
    • P270-Do not eat, drink or smoke when using this product.
    • P273-Avoid release to the environment.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P330-Rinse mouth.
    • P337+P313-If eye irritation persists: Get medical advice/attention.
    • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
    • P403+P235-Store in a well-ventilated place. Keep cool.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contains : 2-Mercaptoethanol, Thiocyanic acid, ethanol