Learn More
Ansell Picosoft DG Light Duty Polyamide Gloves with PVC-Dotted Palm
Product Code. 16895320
Click to view available options
Size:
6
7
8
9
10
11
Gloves are designed for light, delicate applications in dry environments and feature dipped fingertips with PVC dots on the palm for enhanced grip capabilities.
  • Enhanced grip capabilities with dipped fingertips and a PVC-dotted palm
  • Ideal for light and delicate applications in dry environments
  • Ensures tactility and dexterity for precise and safe handling
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material PVC Dots
Color Gray
Hand Specific
Material Polyamide
Size 6
EN388 Standard 2121A
Glove Cuff Style Knit
Series Picosoft DG